Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ Diamond Blister Pack Refill Pipette Tips


Autoclavable or gamma-ray sterlized

Manufacturer: gilson™  F172513

 View more versions of this product

Catalog No. F172513G

  • / Pack of 960 Each
 

Description & Specifications

Specifications

Sterility Sterile
For Use With (Equipment) Pipetmax
Product Type Refill pipette tips
Sterilization Method Gamma-ray sterilized
Model DS200
Packaging Blister Pack
Autoclavable No
Volume (Metric) 2 to 200μL
Tip Style Filtered

  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Features graduations at varying volumes allowing for quick-checks when aspirating samples
  • Standard or filtered tips
  • Validated for use with 384-well tips: F172211G, F172311G and F17511G