Gilson™ PIPETMAN™ Diamond Blister Pack Refill Pipette Tips
Autoclavable or gamma-ray sterlized
Supplier:  Gilson™ F172511
 View more versions of this product
Catalog No. F172511G
Description
Description
  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Features graduations at varying volumes allowing for quick-checks when aspirating samples
  • Standard or filtered tips
  • Validated for use with 384-well tips: F172211G, F172311G and F17511G
Specifications
Specifications
100 to 1000 μL
Sterile
Not Autoclavable
10.79 in.
Rack, Blister
Gilson Pipetmax
Polypropylene
Non-Filtered
Clear
DS1000
Gamma radiation
Universal
Pipetman Diamond