Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ Diamond Blister Pack Refill Pipette Tips
Autoclavable or gamma-ray sterlized
Manufacturer: gilson™  F172211
 View more versions of this product
Catalog No. F172211G
  • / Pack of 960 Each
Description & Specifications
Specifications
Product Type Refill pipette tips
Sterilization Method Gamma-ray sterilized
Sterility Sterile
Tip Style Standard
Volume (Metric) 0.1 to 20μL
Autoclavable No
Packaging Blister Pack
Model DSL10
  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Features graduations at varying volumes allowing for quick-checks when aspirating samples
  • Standard or filtered tips
  • Validated for use with 384-well tips: F172211G, F172311G and F17511G