Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ EasyPack™ Tips
Sealed with an easy open/close zipper lock. Gilson™ PIPETMAN™ EasyPack™ Tips come in bulk packs with assorted sizes.
Manufacturer: gilson™  F161210
 View more versions of this product
Catalog No. F161210G
  • / Pack of 200 Each
Description & Specifications
Specifications
Volume (Metric) 1 to 10mL
Material Polypropylene
Packaging 4 Bags x 50 Tips
Autoclavable Yes
Tip Style Standard
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface
  • Manufactured in a cleanroom and hence no risk of contamination
  • Full traceability from the mold to the bench guaranteed
  • Volume graduations make it easy to visually check the volume aspirated
  • Requires low tip-mounting force
  • Easy identification with color-coded packaging
  • Bulk packs are sealed with a zipper lock which can easily be opened for quick access and closed using the rapid-sealing strip to prevent contamination and loss of tips
  • Each EasyPack box contains 1000 tips (5 bags of 200 tips or 1 bag of 1000; 4 bags of 50 tips or 3 bags of 334 tips)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.