Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ Diamond™ Pipette Tips - Reload Packs
RELOAD PACK is easiest and quickest way to refill empty TIPACK Box
Manufacturer: gilson™  F167013*
 View more versions of this product
Catalog No. F167023G
  • / Pack of 960 Each
Description & Specifications
Specifications
Product Type Reload Pack
Volume (Metric) 2 to 200μL
Model D200
  • Three times smaller than regular tips box
  • Ecofriendly - uses up to 70% less plastic than regular rack box
  • Color-coded racks to differentiate tip models and indicate compatible Gilson pipette models
  • Autoclaving conditions: Moist heat/121°C/20 minutes/1 bar