Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN Classic™ Pipets
Includes “dry seal” technology for air-tightness. Gilson™ PIPETMAN Classic™ Pipets are individually calibrated and delivered with a performance check report.
Manufacturer: gilson™  F123600
 View more versions of this product
Catalog No. F123600G
  • / Each
Description & Specifications
Specifications
For Use With (Application) Small volume dispensing of aqueous fluids of moderate viscosity and density (molecular biology, immunology)
Autoclavable Yes
Accuracy ±0.1, ±5μL
Model P20
Material Stainless Steel and PVDF
Tip Style D200, DF30ST
Volume (Metric) 2 to 20μL
  • Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
  • Tip holder and tip ejector are fully autoclavable to avoid cross-contamination
  • Each pipet bears engraved identification number for traceability