Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN Classic™ Pipets
Includes “dry seal” technology for air-tightness. Gilson™ PIPETMAN Classic™ Pipets are individually calibrated and delivered with a performance check report.
Manufacturer: gilson™  F123615
 View more versions of this product
Catalog No. F123615G
  • / Each
Description & Specifications
Specifications
Accuracy ±0.35, ±1.8μL
Autoclavable Yes
For Use With (Application) Small volume dispensing of aqueous fluids of moderate viscosity and density (molecular biology, immunology)
Material Stainless Steel and PVDF
Tip Style D200, DF100ST
Model P100
Volume (Metric) 20 to 100μL
  • Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
  • Tip holder and tip ejector are fully autoclavable to avoid cross-contamination
  • Each pipet bears engraved identification number for traceability