Please login to your online account to display your discounted pricing

Gilson™ MICROMAN™ Positive-Displacement Pipets


Ensures accuracy and precision when pipetting problem liquids, and offers full protection against cross contamination. Gilson™ MICROMAN™ Positive-Displacement Pipets are 3 to 4 times better than an air displacement pipet when pipetting viscous or volatile liquids.

Manufacturer: gilson™  F148505

 View more versions of this product

Catalog No. F148505G

  • / Each
 

Description & Specifications

Specifications

Autoclavable Yes
Number of Channels 1
Volume (Metric) Range 50 to 250μL
Model M250
Product Type Positive Displacement Pipette
Volume (Metric) 50 to 250μL
For Use With (Application) Pipetting of viscous and dense liquids
Tip Style C250

  • Piston is located inside the tip or capillary preventing pipet contamination
  • Color-coded push-button allows for easy identification of different models
  • Dual function push-button that also ejects capillary pistons
  • Comfortable PVDF handle resistant to aggressive chemicals
  • Accurately measure high vapor pressure liquids without leakage or degradation of the tip
  • M10 and M100 models are fully autoclavable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.