Please login to your online account to display your discounted pricing

Gilson™ MICROMAN™ Positive-Displacement Pipets

Ensures accuracy and precision when pipetting problem liquids, and offers full protection against cross contamination. Gilson™ MICROMAN™ Positive-Displacement Pipets are 3 to 4 times better than an air displacement pipet when pipetting viscous or volatile liquids.

$777.43 - $777.43

Specifications

Product Type Positive Displacement Pipette
Number of Channels 1
Autoclavable Yes
For Use With (Application) Pipetting of viscous and dense liquids
View More Specs
Products
Catalog Number Mfr. No. Tip Style Model Volume (Metric) Price Quantity    

F148501G

 
gilson™
F148501
C10 M10 1 to 10μL Each for $777.43

F148502G

 
gilson™
F148502
C25 M25 30 to 25μL Each for $777.43

F148503G

 
gilson™
F148503
C50 M50 20 to 50μL Each for $777.43

F148504G

 
gilson™
F148504
C100 M100 10 to 100μL Each for $777.43

F148505G

 
gilson™
F148505
C250 M250 50 to 250μL Each for $777.43

F148506G

 
gilson™
F148506
C1000 M1000 100 to 1000μL Each for $777.43
Description & Specifications

Specifications

Product Type Positive Displacement Pipette
Number of Channels 1
Autoclavable Yes
For Use With (Application) Pipetting of viscous and dense liquids

  • Piston is located inside the tip or capillary preventing pipet contamination
  • Color-coded push-button allows for easy identification of different models
  • Dual function push-button that also ejects capillary pistons
  • Comfortable PVDF handle resistant to aggressive chemicals
  • Accurately measure high vapor pressure liquids without leakage or degradation of the tip
  • M10 and M100 models are fully autoclavable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.