Please login to your online account to display your discounted pricing
Gilson™ MICROMAN™ Capillary Pistons
Made specifically for Gilson™ MICROMAN™ E pipets.Gilson™ MICROMAN™ E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs perfectly.
Manufacturer: gilson™  F148312
 View more versions of this product
Catalog No. F148312G
  • / Pack of 960 Each
Description & Specifications
Specifications
For Use With (Equipment) MICROMAN pipet
Product Type Capillary Piston
Material Polypropylene
Sterility Nonsterile
Model CP10
Volume (Metric) 1 to 10μL
  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for Gilson™ MICROMAN™ E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs perfectly.