Please login to your online account to display your discounted pricing

Gilson™ EXPERT™ Universal TOWERPACK™ Racked Pipette Tips Reload System


Be green, save space: snaps tips into place in seconds with just one hand

Manufacturer: gilson™  F1736052

 View more versions of this product

Catalog No. F1736052G

  • / Pack
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Length (English) 2.28 in.
For Use With (Equipment) Universal
Material Inert Polymer
Product Type Expert™ Tip
Length (Metric) 5.8cm
Sterility Nonsterile
Packaging Tower Pack Racked
Graduations 2, 5, 10μL
Model E300
Certifications/Compliance DNase/RNase-free, Non-pyrogenic, Human gDNA and PCR inhibition-free
Color Green
Tip Style Extended Length
Volume (Metric) 1 to 300μL
Autoclavable No

Reload system gives you easy access to 10 layers of tips that easily snap into the Tipack boxes.

  • Fit leading pipetter brands
  • Allow you to stock one brand of tip with practical packaging options so you can save time, money and bench space
  • Produced in a sterile, clean-room environment using the purest raw materials and state-of-the-art manufacturing
  • Offer consistent performance
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and pyrogens
  • Feature an inert polymer hydrophobic surface to minimize sample binding when dispensing
  • Clear packaging allows easy tip identification
  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipetters
  • Feature graduations at varying volumes allowing for quick checks when aspirating samples
  • Economical and easy to use

  • TowerPack: 960 Tips/Pk.
  • E1000 XL Towerpack: 768 Tips/Pk.