Please login to your online account to display your discounted pricing

Gilson™ EXPERT™ Universal TOWERPACK™ Racked Pipette Tips Reload Kit


Be green, save space: snaps tips into place in seconds with just one hand with tips plus Tipack boxes

Manufacturer: gilson™  F1735061

 View more versions of this product

Catalog No. F1735061G

  • / Pack
 

Description & Specifications

Specifications

Product Type Expert™ Tip
Material Inert Polymer
Length (English) 4 in.
Length (Metric) 10.2cm
For Use With (Equipment) Universal
Sterility Nonsterile
Graduations 200, 500, 1000μL
Model E1000 XL
Tip Style Extended Length
Packaging Tower Pack Racked
Certifications/Compliance DNase/RNase-free, Non-pyrogenic, Human gDNA and PCR inhibition-free
Volume (Metric) 50 to 1250μL
Color Blue

Gives you easy access to 10 layers of tips that easily snap into the Tipack boxes.

  • Fit leading pipetter brands
  • Allow you to stock one brand of tip with practical packaging options so you can save time, money and bench space
  • Produced in a sterile, clean-room environment using the purest raw materials and state-of-the-art manufacturing
  • Offer consistent performance
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and pyrogens.
  • Feature an inert polymer hydrophobic surface to minimize sample binding when dispensing
  • Clear packaging allows easy tip identification
  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipetters
  • Feature graduations at varying volumes allowing for quick checks when aspirating samples
  • Economical and easy to use

  • TowerPack Kits: 960 Tips/Pk., 10 empty Tipack boxes
  • E1000 XL TowerPack Kit: 768 Tips/Pk., 8 empty Tipack Boxes