Gilson™ Tip Empty Rack For Pipet Tips
Use with loose Gilson Diamond tips. Gilson™ Tip Empty Rack For Pipette Tips and are colour coded for ease of use.
Manufacturer:  Gilson™ F171502
 View more versions of this product
Catalog No. F171502G
Specifications
Description
Rack, Empty; Gilson PIPETMAN; For 100 - 1000uL D1000 pipet tips; Fully autoclavable; For manual loading only; Made up of pure polypropylene; Set of 2
  • Fully autoclavable
  • Made of polypropylene
Description & Specifications
Specifications
Rack, Empty
Yes
100 to 1000μL D1000 pipet tips
Polypropylene