Please login to your online account to display your discounted pricing
Gilson™ DistriTips™ Syringes
Use of Gilson™’s pipetting system guarantees accuracy and precision. Gilson™ DistriTips™ Syringes have an aliquot volume range from 1µL to 1.25mL
Manufacturer: gilson™  F164110
 View more versions of this product
Catalog No. F164110G
  • / Pack of 50 Each
Description & Specifications
Specifications
For Use With (Application) Viscous, volatile solutions
Model 1250μL Mini
Type DistriTip™ Syringe
Volume (Metric) 10μL to 125μL
Quantity 50 Pack
  • With level indicator
  • Available in standard or sterilized (ST) form, in boxes of 50
  • Certified free of RNase, DNase, DNA, RNA, and Protease
  • Free of dyes, so no risk of contamination from their components
  • Only three syringes are needed to cover 1μ to 1.25mL
  • More than 100 aliquots can be dispensed from each syringe
  • Ideal for precise aliquots
  • Can be used with viscous and volatile solutions
  • Individually wrapped
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.