Please login to your online account to display your discounted pricing

Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Reagent Reservoirs


Ideal for dispensing reagents with multichannel pipettes. Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Reagent Reservoirs' unique shape allows for efficient and nearly complete sample withdrawal.

Manufacturer: gilson™  F267670

 View more versions of this product

Catalog No. F267670G

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Volume (Metric) 50mL
For Use With (Equipment) Pipetteman™ complete pipetting system
Material Polystyrene
Volume (Metric) 50mL
Color White
Color White
Product Type Reagent Reservoir

 

  • Angled sides and V-shaped bottoms
  • Pour-off spouts on all corners
  • Graduations on inside walls at middle volume