Gilson™ Accessory for PIPETMAN™ Complete Pipetting System - Reagent Reservoir
Ideal for dispensing reagents with multichannel pipettes. Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Reagent Reservoirs' unique shape allows for efficient and nearly complete sample withdrawal.
Supplier: Gilson™ F267660
Description
- Angled sides and V-shaped bottoms
- Pour-off spouts on all corners
- Graduations on inside walls at middle volume
Specifications
White | |
Polystyrene | |
25 mL | |
100 Pk. |
All Pipettes with Tips Mounted | |
Reservoir | |
PIPETMAN™ Complete Pipetting System |