Please login to your online account to display your discounted pricing

Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Coloris™ Colored Identifi cation Clips


GLP-friendly, brightly colored clips made of polyacetal are used for identifying Pipetteman™ and Neo™ pipets

Manufacturer: gilson™  F161302

 View more versions of this product

Catalog No. F161302G

  • / Pack of 10 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Pipetteman™ and Neo™ pipettes
Product Type Coloris™ identification clip
Color Red
Material Polyacetal
Color Red

  • Brightly colored clips
  • Made of polyacetal
  • Used for identifying Pipetteman and Neo pipettes