Gilmont™ Gilmont™ Micrometer Syringes and Burets
Experience high precision with economy of cost when using Gilmont™ Micrometer Syringes.
$915.74 - $915.74
Products 1
Catalog Number Mfr. No. Type Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Type Volume (Metric) Price Quantity  
15-078-150
View Documents
Gilmont™
0784000
Syringe 0.2 mL
Each for $915.74
Description
Description
Experience high precision with economy of cost when using Thermo Scientific™ Gilmont Micrometer Syringes.
  • 0.5% accuracy
  • Liquid comes in contact with only glass, VITON™ and PTFE™
  • VITON O-ring gives a vacuum tight seal
  • All other plastic parts of polypropylene--instrument may be autoclaved
  • Available in two sizes, 0.2 mL in 0.0002 mL div. and 2.0 mL in 0.002 mL div.
  • Fills and delivers liquid with micrometer control
  • Simple to clean and assemble
  • SDS