Please login to your online account to display your discounted pricing

Giemsa Stain Solution, Hematology Stain, EMD Millipore


Quantity 500mL
Packaging Poly Bottle

Manufacturer: emd millipore corporation  R0305574

 View more versions of this product

Catalog No. MR0305574

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Chemical Name or Material Giemsa Stain Solution
Name Note Hematology Stain
Packaging Poly Bottle
Synonym Hematology Stain

Specifications

Chemical Name or Material Giemsa Stain Solution
Name Note Hematology Stain
Synonym Hematology Stain