missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Giemsa Stain Solution, Hematology Stain, MilliporeSigma™
$372.71 - $581.97
Chemical Identifiers
CAS | 56-81-5 |
---|---|
Molecular Formula | C3H8O3 |
Molecular Weight (g/mol) | 92.09 |
MDL Number | MFCD00004722 |
InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
Synonym | Hematology Stain |
PubChem CID | 753 |
ChEBI | CHEBI:17754 |
IUPAC Name | propane-1,2,3-triol |
SMILES | OCC(O)CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
MR0305574
|
Emd Millipore Corporation
R0305574 |
500mL | Poly Bottle |
Each for $372.71
|
|
MR0305576
|
Emd Millipore Corporation
R0305576 |
1L | Poly Bottle |
Each for $438.56
|
|
MR0305582
|
Emd Millipore Corporation
R0305582 |
4L | Poly Bottle |
Each for $581.97
|
|
Chemical Identifiers
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
Hematology Stain | |
CHEBI:17754 | |
OCC(O)CO |
Specifications
56-81-5 | |
MFCD00004722 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
Poly Bottle | |
Giemsa Stain Solution, Hematology Stain |
C3H8O3 | |
Hematology Stain | |
OCC(O)CO | |
92.09 | |
CHEBI:17754 | |
500mL |