missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Giemsa Stain, Reagent Grade, LabChem™

$168.80 - $168.80

Chemical Identifiers

CAS 51811-82-6
CAS Max % 100
Linear Formula C14H14ClN3S
Formula Weight 291.8
Chemical Name or Material Giemsa Stain
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC148357 Lab Chem
LC148357
Each for $168.80
Specifications Description & Specifications

Chemical Identifiers

51811-82-6
C14H14ClN3S
Giemsa Stain
100
291.8
Reagent Grade

Specifications

51811-82-6
Passes Test
Amber Glass
291.8
Hydrogen chloride; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Greenish-gold
Reagent Grade
100
Powder
C14H14ClN3S
Soluble in water
300°C
Giemsa Stain

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 257-438-2

Recommended Storage : Room Temperature