Giemsa Stain (Powder/Certified Biological Stain), Fisher Chemical™
Manufacturer:  Fisher Chemical G1465
 View more versions of this product
Catalog No. G146-5
Description
Description
Specifications
Specifications
Giemsa Stain
Pass Test
MFCD00012112,MFCD00081642
NALREUIWICQLPS-UHFFFAOYSA-N
7-amino-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
13735
Certified Biological Stain
300°C
5g
51811-82-6
C14H14ClN3S
Pass Test
[Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1
291.80
Solid
Glass Bottle
Green
Chemical Identifiers
51811-82-6
291.80
NALREUIWICQLPS-UHFFFAOYSA-N
7-amino-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
C14H14ClN3S
MFCD00012112,MFCD00081642
13735
[Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1