missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Giemsa Stain (Powder/Certified Biological Stain), Fisher Chemical™
Manufacturer: Fisher Chemical G14610
Specifications
Giemsa Stain | |
Pass Test | |
MFCD00012112,MFCD00081642 | |
NALREUIWICQLPS-UHFFFAOYSA-N | |
7-amino-N,N-dimethyl-3H-phenothiazin-3-iminium chloride | |
13735 | |
Certified Biological Stain | |
300°C | |
10g |
51811-82-6 | |
C14H14ClN3S | |
Pass Test | |
[Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1 | |
291.80 | |
Solid | |
Glass Bottle | |
Green |
Chemical Identifiers
51811-82-6 | |
291.80 | |
NALREUIWICQLPS-UHFFFAOYSA-N | |
7-amino-N,N-dimethyl-3H-phenothiazin-3-iminium chloride |
C14H14ClN3S | |
MFCD00012112,MFCD00081642 | |
13735 | |
[Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1 |
Safety and Handling
