missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Giemsa Stain (Powder/Certified Biological Stain), Fisher Chemical

$145.54 - $199.92

Chemical Identifiers

CAS 51811-82-6
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.797
MDL Number MFCD00081642
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
G146-5 View Documents Fisher Chemical
G1465
5g Glass Bottle Each for $145.54
Case of 6 Each for $873.24
 
G146-10 View Documents Fisher Chemical
G14610
10g Glass Bottle Each for $199.92
Case of 6 Each for $1,199.52
 
Specifications

Chemical Identifiers

51811-82-6
291.797
NALREUIWICQLPS-UHFFFAOYSA-N
(7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
C14H14ClN3S
MFCD00081642
13735
C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-]

Specifications

51811-82-6
C14H14ClN3S
Pass Test
C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-]
291.797
Solid
Glass Bottle
Green
Giemsa Stain
Pass Test
MFCD00081642
NALREUIWICQLPS-UHFFFAOYSA-N
(7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
13735
Certified Biological Stain
300°C
5g
Safety and Handling

Safety and Handling