Please login to your online account to display your discounted pricing

Giemsa's azur-eosin-methylene blue solution, For Microscopy, EMD Millipore™


Manufacturer: emd millipore corporation  1092040100

 View more versions of this product

Catalog No. M1092040100

  • / Each
 

Description & Specifications

Chemical Identifiers

DOT Information DOT Class: 3 (6.1), Packaging Group: II
Chemical Name or Material Giemsa's azur-eosin-methylene blue solution
Physical Form Liquid
UN Number UN 1230
Health Hazard 1 H225: Highly flammable liquid and vapour.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
H370: Causes damage to organs
Odor Smells like methanol
Packaging Glass bottle
Appearance Blue liquid

Specifications

Chemical Name or Material Giemsa's azur-eosin-methylene blue solution
UN Number UN 1230
Accessories

H225: Highly flammable liquid and vapour.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
H370: Causes damage to organs