Please login to your online account to display your discounted pricing

Giemsa's azur-eosin-methylene blue solution, For Microscopy, EMD Millipore™

$133.66 - $6,231.43

Chemical Identifiers

Chemical Name or Material Giemsa's azur-eosin-methylene blue solution
UN Number UN 1230
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

M1092040100

 
emd millipore corporation
1092040100
Each for $133.66

M1092040500

 
emd millipore corporation
1092040500
Each for $208.89

M1092041000

 
emd millipore corporation
1092041000
Each for $374.14

M1092042500

 
emd millipore corporation
1092042500
Each for $623.71

M1092049025

 
emd millipore corporation
1092049025
Each for $6,231.43
Description & Specifications

Specifications

DOT Information DOT Class: 3 (6.1), Packaging Group: II
Physical Form Liquid
Health Hazard 1 H225: Highly flammable liquid and vapour.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
H370: Causes damage to organs
UN Number UN 1230
Odor Smells like methanol
Packaging Glass bottle
Appearance Blue liquid
Chemical Name or Material Giemsa's azur-eosin-methylene blue solution

For microscopy

Safety and Handling

H225: Highly flammable liquid and vapour.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
H370: Causes damage to organs