missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ VEGF Recombinant Rat Protein DFS Item

Recombinant VEGF is a bioactive protein intended for use in cell culture applications

$358.00 - $4,560.00

Specifications

Purity 95%
Purification Method Exchange Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias VEGF
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRG0111 Gibco™
LSPRG0111
100μg Each for $1,338.00
PRG0113 Gibco™
LSPRG0113
1mg Each for $4,560.00
PRG0114 Gibco™
LSPRG0114
10μg Each for $358.00
PRG0115 Gibco™
LSPRG0115
25μg Each for $778.00
Specifications

Description

VEGF exerts angiogenic, mitogenic and vascular permeability-enhancing activities specific for endothelial cells. VEGF has also been shown to be chemotactic for monocytes and osteoblasts.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

95%
<0.1ng/μg
VEGF
Exchange Chromatography
Lyophilized
83785