Please login to your online account to display your discounted pricing

Gibco™ VEGF Recombinant Rat Protein DFS Item


Recombinant VEGF is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPRG0113

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PRG0113

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Angiogenesis
Storage Requirements Store at 2° to 8°C.
Purity 95%
Quantity 1mg
Format Lyophilized
Endotoxin Concentration <0.1ng/μg
Source Rat
Protein Family Growth Factors & Receptors
Molecular Weight 38.4kDa
Purification Method Exchange Chromatography
Without Additives Carrier Protein
Protein Subtype VEGF (Vascular Endothelial Growth Factor)
Gene Alias VEGF
Structural Form Recombinant/Ligand
Validated Application Cell Culture
Expression System E. coli

VEGF exerts angiogenic, mitogenic and vascular permeability-enhancing activities specific for endothelial cells. VEGF has also been shown to be chemotactic for monocytes and osteoblasts.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.