Please login to your online account to display your discounted pricing

Gibco™ VEGF Recombinant Mouse Protein DFS Item


Recombinant Mouse Vascular Endothelial Cell Growth Factor (VEGF) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMG0111

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PMG0111

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 38.8kDa
Quantity 100μg
Protein Family Growth Factors & Receptors
Expression System E. coli
Purification Method Exchange Chromatography
Purity 95%
Source Mouse
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype VEGF (Vascular Endothelial Growth Factor)
Endotoxin Concentration <0.1ng/μg
Gene Alias VEGF
Validated Application Cell Culture
For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Angiogenesis
Format Lyophilized
Gene ID (Entrez) 22339

VEGF exerts angiogenic, mitogenic and vascular permeability-enhancing activities specific for endothelial cells. VEGF has also been shown to be chemotactic for monocytes and osteoblasts.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human umbilical vein cells (HUVEC)

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.