Please login to your online account to display your discounted pricing

Gibco™ TPO Recombinant Mouse Protein DFS Item


Recombinant TPO is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC1144

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PMC1144

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 17kDa
Quantity 10μg
Storage Requirements Store at 2° to 8°C.
Format Lyophilized
Purification Method Sequential Chromatography
Purity 95%
Source Mouse
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype TPO
Endotoxin Concentration <0.1ng/μg
Gene Alias TPO
Validated Application Cell Culture
Protein Family Growth Factors & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
Gene ID (Entrez) 22018

TPO is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.