Please login to your online account to display your discounted pricing

Gibco™ TNF Recombinant Rat Protein DFS Item


Recombinant TNF is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPRC3014

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PRC3014

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 17kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 10μg
Source Rat
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype TNF (Tumor Necrosis Factor)
Endotoxin Concentration <0.1ng/μg
Gene Alias TNF-α
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Cell Signaling
Format Lyophilized
Gene ID (Entrez) 24835

TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.