Please login to your online account to display your discounted pricing

Gibco™ TNF Recombinant Rat Protein DFS Item


Recombinant TNF is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPRC3013

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PRC3013

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Storage Requirements Store at 2° to 8°C.
Structural Form Recombinant/Ligand
Gene ID (Entrez) 24835
Without Additives Carrier Protein
Validated Application Cell Culture
Expression System E. coli
Protein Subtype TNF (Tumor Necrosis Factor)
Endotoxin Concentration <0.1ng/μg
Purity 95%
Purification Method Sequential Chromatography
Quantity 1mg
Gene Alias TNF-α
For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Cell Signaling
Molecular Weight 17kDa
Source Rat
Protein Family Cytokines & Receptors

TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.