Please login to your online account to display your discounted pricing

Gibco™ TNF Recombinant Mouse Protein DFS Item


Recombinant Mouse Tumor Necrosis Factor-α (TNF) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC3014

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PMC3014

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture; Cell Signaling
Quantity 10μg
Source Mouse
Storage Requirements Store at 2° to 8°C.
Purification Method Sequential Chromatography
Gene ID (Entrez) 21926
Structural Form Recombinant/Ligand
Endotoxin Concentration <0.1ng/μg
Expression System E. coli
Protein Subtype TNF (Tumor Necrosis Factor)
Gene Alias TNF-α
Protein Family Cytokines & Receptors
Molecular Weight 17.5kDa
Validated Application Cell Culture
Purity 95%
Without Additives Carrier Protein
Format Lyophilized

TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent cytotoxicity of L929 cells

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.