Please login to your online account to display your discounted pricing

Gibco™ TIMP1 Recombinant Human Protein DFS Item

Recombinant TIMP1 is a bioactive protein intended for use in cell culture applications

$285.60 - $6,720.00

Specifications

Gene ID (Entrez) 443331, 73858576
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC8021

DFS Item
 
gibco™
PHC8021
100μg Each for $1,190.70

PHC8023

DFS Item
 
gibco™
PHC8023
1mg Each for $6,720.00

PHC8024

DFS Item
 
gibco™
PHC8024
10μg Each for $285.60
Description & Specifications

Specifications

Gene ID (Entrez) 443331, 73858576

TIMP1 is an inhibitor of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix. In addition to its inhibitory role against most of the knownmMPs, the encoded protein is able to promote cell proliferation in a wide range of cell types, and may also have an anti-apoptotic function.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.