Please login to your online account to display your discounted pricing

Gibco™ TIMP1 Recombinant Human Protein DFS Item


Recombinant TIMP1 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC8024

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC8024

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 2° to 8°C.
Source Human
Quantity 10μg
Validated Application Cell Culture
For Use With (Application) Cell Culture; Mammalian Cell Culture
Gene ID (Entrez) 443331, 73858576
Protein Subtype TIMP
Protein Family Proteases, Peptidases & Inhibitors
Structural Form Recombinant

TIMP1 is an inhibitor of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix. In addition to its inhibitory role against most of the knownmMPs, the encoded protein is able to promote cell proliferation in a wide range of cell types, and may also have an anti-apoptotic function.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.