Please login to your online account to display your discounted pricing

Gibco™ TIMP1 Recombinant Human Protein DFS Item


Recombinant TIMP1 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC8021

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC8021

  • / Each
 

Description & Specifications

Specifications

Quantity 100μg
Source Human
Storage Requirements Store at 2° to 8°C.
Structural Form Recombinant
Protein Subtype TIMP
Validated Application Cell Culture
Protein Family Proteases, Peptidases & Inhibitors
For Use With (Application) Cell Culture; Mammalian Cell Culture
Gene ID (Entrez) 443331, 73858576

TIMP1 is an inhibitor of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix. In addition to its inhibitory role against most of the knownmMPs, the encoded protein is able to promote cell proliferation in a wide range of cell types, and may also have an anti-apoptotic function.

Shipping Condition: Wet Ice.