missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ TGFB3 Recombinant Human Protein DFS Item

Recombinant Human Transforming Growth Factor β 3 (TGFB3) is a bioactive protein intended for use in cell culture applications

$256.00 - $8,260.00

Specifications

Purity 98%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Liquid
Gene Alias TGF-β3
View More Specs

Disclaimers:

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHG9302 Gibco™
LSPHG9302
250μg Each for $8,260.00
PHG9305 Gibco™
LSPHG9305
5μg Each for $256.00
Specifications

Description

TGFB3 is involved in epithelial and endothelial cell proliferation and differentiation during development.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined in a toxicity assay using WHO Standard 98/608 for direct comparison

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

98%
<0.1ng/μg
TGF-β3
Sequential Chromatography
Liquid
7043