Please login to your online account to display your discounted pricing

Gibco™ TGFB2 Recombinant Human Protein DFS Item


Recombinant Human Transforming Growth Factor β 2 (TGFB2) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHG9114

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PHG9114

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Embryonic Stem Cell Culture; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture; Differentiation & Development
Structural Form Recombinant/Ligand
Purity 98%
Format Lyophilized
Molecular Weight 25kDa
Endotoxin Concentration <0.1ng/μg
Source Human
Gene ID (Entrez) 7042
Purification Method Sequential Chromatography
Gene Alias TGF-β2
Expression System Baculovirus
Protein Family Growth Factors & Receptors
Protein Subtype TGFs (Transforming Growth Factors)
Storage Requirements Store at -5° to -30°C.
Validated Application Cell Culture
Quantity 5μg

TGFB2 binds the transforming growth factor-β receptor and plays a role in regulation of cell growth and proliferation. It may be involved in mesenchymal-epithelial cell interactions during development.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by its ability to inhibit the mouse IL-4 dependent proliferation of mouse HT-2 cells

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.