Please login to your online account to display your discounted pricing

Gibco™ Sonic Hedgehog C25II Recombinant Mouse Protein DFS Item

Recombinant Sonic Hedgehog C25II is a bioactive protein intended for use in cell culture applications

$271.00 - $8,420.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias Sonic Hedgehog C25II
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PMC8031

DFS Item
 
gibco™
LSPMC8031
100μg Each for $1,276.00

PMC8033

DFS Item
 
gibco™
LSPMC8033
1mg Each for $8,420.00

PMC8034

DFS Item
 
gibco™
LSPMC8034
10μg Each for $271.00

PMC8035

DFS Item
 
gibco™
LSPMC8035
25μg Each for $373.80
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias Sonic Hedgehog C25II
Gene ID (Entrez) 6516263
Accession Number BC063088

Sonic Hedgehog is instrumental in patterning the early embryo. It has been implicated as the key inductive signal in patterning of the ventral neural tube, the anterior-posterior limb axis, and the ventral somites.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.