Please login to your online account to display your discounted pricing

Sf-900™ III SFM DFS Item


Low-hydrolysate, serum-free, protein-free, animal origin-free

Manufacturer: life technologies  LS12658035

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12658035

  • / Each
 

Description & Specifications

Specifications

Quantity 10L
For Use With (Application) Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production
Storage Requirements Storage conditions: 2° to 8°C. Protect from light. Shelf life: 18 months from date of manufacture
Additive L-Glutamine
Format Liquid
Species S. frugiperda, Spodoptera frugiperda
Cell Line Sf21, Sf9

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.