Please login to your online account to display your discounted pricing

Sf-900™ II SFM DFS Item


Optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells

Manufacturer: life technologies  LS10902088

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10902088

  • / Each
 

Description & Specifications

Specifications

Format Liquid
Quantity 1000mL
Species S. frugiperda, Spodoptera frugiperda
For Use With (Application) Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production
Additive L-Glutamine
Cell Line Sf21, Sf9
Storage Requirements Storage conditions: 2°C to 8°C. Protect from light. Shelf life: 12 months from date of manufacture

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.