Please login to your online account to display your discounted pricing

Gibco™ SDF1B Recombinant Human Protein DFS Item


Recombinant SDF1B is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC1364

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC1364

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 8.7kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 10μg
Source Human
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype SDFs (Stromal Cell-Derived Factors)
Endotoxin Concentration <0.1ng/μg
Gene Alias SDF-1β
Validated Application Cell Culture
Protein Family Chemokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture
Format Lyophilized
Gene ID (Entrez) 6387

SDF1B is a small cytokine that belongs to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.