Please login to your online account to display your discounted pricing

Gibco™ SDF1A Recombinant Human Protein DFS Item

Recombinant SDF1A is a bioactive protein intended for use in cell culture applications

$257.00 - $1,994.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias SDF-1α
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC1341

DFS Item
 
gibco™
LSPHC1341
100μg Each for $1,994.00

PHC1344

DFS Item
 
gibco™
LSPHC1344
10μg Each for $257.00

PHC1346

DFS Item
 
gibco™
LSPHC1346
25μg Each for $630.00
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias SDF-1α
Gene ID (Entrez) 6387

SDF1A is a small cytokine that belongs to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.