Please login to your online account to display your discounted pricing

Gibco™ SDF1A Recombinant Human Protein DFS Item


Recombinant SDF1A is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC1346

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC1346

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 8kDa
Gene ID (Entrez) 6387
Storage Requirements Store at 2° to 8°C.
Purification Method Sequential Chromatography
Source Human
Quantity 25μg
Gene Alias SDF-1α
Expression System E. coli
Purity 95%
Protein Family Chemokines & Receptors
Protein Subtype SDFs (Stromal Cell-Derived Factors)
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Validated Application Cell Culture
Format Lyophilized
For Use With (Application) Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture
Endotoxin Concentration <0.1ng/μg

SDF1A is a small cytokine that belongs to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.