missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ SDF1A Recombinant Human Protein DFS Item

Recombinant SDF1A is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPHC1344

 View more versions of this product

Catalog No. PHC1344



Description

Description

SDF1A is a small cytokine that belongs to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture
6387
Sequential Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
8kDa
95%
Human
Carrier Protein
SDFs (Stromal Cell-Derived Factors)
SDF-1α
Chemokines & Receptors
Documents

For Research Use Only. Not for use in diagnostic procedures.