Please login to your online account to display your discounted pricing

Gibco™ SCF (C-Kit Ligand) Recombinant Mouse Protein DFS Item

Recombinant Mouse Stem Cell Factor (SCF, C-Kit Ligand) is a bioactive protein intended for use in cell culture applications

$358.00 - $3,455.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Liquid
Gene Alias SCF, c-kit Ligand
View More Specs

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PMC2113L

DFS Item
 
gibco™
LSPMC2113L
1mg Each for $3,455.00

PMC2111

DFS Item
 
gibco™
LSPMC2111
100μg Each for $976.00

PMC2114

DFS Item
 
gibco™
LSPMC2114
10μg Each for $358.00

PMC2115

DFS Item
 
gibco™
LSPMC2115
25μg Each for $654.00
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Liquid
Gene Alias SCF, c-kit Ligand

SCF is a pleiotropic factor that acts in utero in germ cell and neural cell development, and hematopoiesis, all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human MO-7e cells

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.