Please login to your online account to display your discounted pricing

Gibco™ SCF (C-Kit Ligand) Recombinant Mouse Protein DFS Item


Recombinant Mouse Stem Cell Factor (SCF, C-Kit Ligand) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC2115

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PMC2115

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Embryonic Stem Cell Culture; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture & Development
Molecular Weight 18.5kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 25μg
Source Mouse
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype Other Proteins
Endotoxin Concentration <0.1ng/μg
Gene Alias SCF, c-kit Ligand
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System E. coli
Format Lyophilized
Gene ID (Entrez) 17311

SCF is a pleiotropic factor that acts in utero in germ cell and neural cell development, and hematopoiesis, all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human MO-7e cells

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.