missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ RANTES Recombinant Human Protein DFS Item

Recombinant RANTES is a bioactive protein intended for use in cell culture applications

$266.00 - $4,400.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias RANTES
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC1051 Gibco™
LSPHC1051
100μg Each for $1,038.00
PHC1053 Gibco™
LSPHC1053
1mg Each for $4,400.00
PHC1054 Gibco™
LSPHC1054
10μg Each for $266.00
Specifications

Description

RANTES functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. RANTES is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.

Shipping Condition: Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

95%
<0.1ng/μg
RANTES
Sequential Chromatography
Lyophilized
6352