missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ RANTES Recombinant Human Protein DFS Item

Recombinant RANTES is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPHC1054

 View more versions of this product

Catalog No. PHC1054


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Gibco; RANTES Recombinant Human Protein; 10ug; Source: Human; Purity: 95%

RANTES functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. RANTES is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.

Shipping Condition: Wet Ice.

Description & Specifications

Specifications

Cell Culture; Mammalian Cell Culture
6352
Sequential Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
7.8kDa
95%
Human
Carrier Protein
Rantes
RANTES
Chemokines & Receptors