missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ PFHM-II Protein-Free Hybridoma Medium DFS Item

Serum-free, protein-free medium specifically developed for the ability to support the growth of hybridomas for antibody production

$122.00 - $330.00

Specifications

Format Liquid
Packaging Sustainable packaging
Storage Requirements Storage: 2-8°C. Shelf life: 24 months from date of manufacture
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12040077 Life Technologies
LS12040077
1000mL Each for $122.00
23600042 Life Technologies
LS23600042
1 x 10L Each for $330.00
Specifications

Description

  • Ability to support suspension and stationary hybridomas and myelomas
  • Complete, ready-to-use, serum-free, protein-free formulation
  • Ability to support suspension and stationary hybridomas and myelomas
  • Suitable for culture of recombinant myeloma lines as well as traditional hybridomas, Gibco PFHM II provides high cell densities and IgG production
  • Cholesterol-dependent cultures such as NS0 require the addition of 250x Cholesterol Lipid Concentrate
  • Sustainable packaging

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco PFHM II is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Description & Specifications

Specifications

Liquid
Sustainable packaging
Storage: 2-8°C. Shelf life: 24 months from date of manufacture